siRNA Details
Gene Symbol: Wipf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289722.1 | NP_001276651.1 | 7 | 1610 | ||
NM_001289723.1 | NP_001276652.1 | 7 | 1575 | |||
NM_153138.4 | NP_694778.1 | 7 | 1605 | |||
GenBank mRNA | AJ437262.1 | - | 6 | 1406 | ||
AK031046.1 | - | 7 | 1572 | |||
AK156547.1 | - | 7 | 1610 | |||
AK168092.1 | - | 3 | 759 | |||
BC049788.1 | - | 7 | 1528 | 6514406 |
