siRNA Details
Gene Symbol: Fcgbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122603.1 | NP_001116075.1 | 12 | 4452 | ||
GenBank mRNA | AJ011416.1 | - | 6 | 1790 | ||
AK040441.1 | - | 2 | 1156 | |||
BC030871.1 | - | 1 | 301 | |||
BC039286.1 | - | 3 | 880 |
