siRNA Details
Gene Symbol: Taok1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144825.2 | NP_659074.2 | 20 | 3422 | ||
GenBank mRNA | AK173156.1 | - | 4 | 756 | ||
BC016522.1 | - | 8 | 1416 | |||
BC034906.1 | - | 2 | 265 | |||
BC047271.1 | - | 19 | 2769 | |||
BC151012.1 | - | 20 | 3216 | 9087925 | ||
BC151015.1 | - | 20 | 3216 | 9087928 | ||
BC151016.1 | - | 20 | 3215 | 9087929 |
