siRNA Details
Gene Symbol: Fam208a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114879.1 | NP_001108351.1 | 15 | 2746 | ||
NM_028945.1 | NP_083221.1 | 15 | 2746 | |||
GenBank mRNA | AK028620.1 | - | 2 | 497 | ||
AK044563.1 | - | 15 | 2747 | |||
BC037390.1 | - | 1 | 237 |
