siRNA Details
Gene Symbol: Fam208a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114879.1 | NP_001108351.1 | 23 | 4540 | ||
NM_028945.1 | NP_083221.1 | 23 | 4540 | |||
GenBank mRNA | AK028620.1 | - | 10 | 2290 | ||
AK173099.1 | - | 3 | 275 | |||
BC037390.1 | - | 9 | 2031 |
