siRNA Details
Gene Symbol: Tpd52l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009413.1 | NP_033439.1 | 5, 6 | 621 | ||
GenBank mRNA | AF004428.1 | - | 5, 6 | 621 | ||
AK182630.1 | - | 2, 3 | 83 | |||
BC141068.1 | - | 6, 7 | 523 | 9055680 | ||
BC145321.1 | - | 6, 7 | 523 | 9053876 |
