siRNA Details
Gene Symbol: Tpd52l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009413.1 | NP_033439.1 | 6 | 654 | ||
GenBank mRNA | AF004428.1 | - | 6 | 654 | ||
AK182630.1 | - | 3 | 116 | |||
BC141068.1 | - | 7 | 556 | 9055680 | ||
BC145321.1 | - | 7 | 556 | 9053876 |
