siRNA Details
Gene Symbol: Tpd52l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009413.1 | NP_033439.1 | 7 | 753 | ||
GenBank mRNA | AF004428.1 | - | 7 | 753 | ||
AK131855.1 | - | 5 | 550 | |||
AK182630.1 | - | 4 | 215 | |||
BC141068.1 | - | 8 | 655 | 9055680 | ||
BC145321.1 | - | 8 | 655 | 9053876 | ||
BC145322.1 | - | 7 | 594 | |||
BC171924.1 | - | 5 | 540 |
