siRNA Details
Gene Symbol: Tsc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039363.2 | NP_001034452.1 | 12, 13 | 1378 | ||
NM_001286713.1 | NP_001273642.1 | 12, 13 | 1378 | |||
NM_001286714.1 | NP_001273643.1 | 12, 13 | 1373 | |||
NM_001286716.1 | NP_001273645.1 | 12, 13 | 1373 | |||
NM_001286718.1 | NP_001273647.1 | 12, 13 | 1378 | |||
NM_001286720.1 | NP_001273649.1 | 12, 13 | 1378 | |||
NM_011647.3 | NP_035777.2 | 12, 13 | 1378 | |||
GenBank mRNA | AF132986.1 | - | 8, 9 | 906 | ||
AK143799.1 | - | 12, 13 | 1379 | |||
AK147016.1 | - | 12, 13 | 1354 | |||
AK147302.1 | - | 12, 13 | 1318 | |||
AK147487.1 | - | 12, 13 | 1347 | |||
AK147874.1 | - | 12, 13 | 1343 | |||
AK147908.1 | - | 12, 13 | 1379 | |||
AK148062.1 | - | 12, 13 | 1370 | |||
AK148072.1 | - | 12, 13 | 1275 | |||
AK170589.1 | - | 12, 13 | 1369 | |||
AK171007.1 | - | 12, 13 | 1347 | |||
AK217801.1 | - | 1, 2 | 127 | |||
AK218975.1 | - | 1, 2 | 127 | |||
BC052449.1 | - | 12, 13 | 1354 | 6409037 | ||
BC060701.1 | - | 12, 13 | 1354 | 6409037 | ||
U37775.1 | - | 11, 12 | 1290 | |||
U39818.1 | - | 12, 13 | 1349 |
