siRNA Details
Gene Symbol: Ube2g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019803.3 | NP_062777.2 | 5 | 403 | ||
GenBank mRNA | AF296657.1 | - | 5 | 384 | ||
AK136365.1 | - | 5 | 390 | |||
AK146554.1 | - | 5 | 408 | |||
AK154464.1 | - | 5 | 424 | |||
BC010321.1 | - | 5 | 403 | 2651046 | ||
U93241.1 | - | 4 | 303 |
