siRNA Details
Gene Symbol: Uros | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA272198.1 | - | 1 | 150 | ||
AK002971.1 | - | 2 | 200 | |||
AK135314.1 | - | 2 | 206 | |||
AK145902.1 | - | 2 | 256 | |||
AK148116.1 | - | 2 | 393 | |||
AK164297.1 | - | 2 | 171 | |||
BC068161.1 | - | 2 | 199 | 30358504 | ||
U04439.1 | - | 1 | 120 | |||
U16216.1 | - | 2 | 261 | |||
U18867.2 | - | 2 | 265 |
