siRNA Details
Gene Symbol: Uros | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA272198.1 | - | 5, 6 | 513 | ||
AK135314.1 | - | 6, 7 | 569 | |||
AK145902.1 | - | 6, 7 | 619 | |||
AK148116.1 | - | 6, 7 | 756 | |||
BC068161.1 | - | 6, 7 | 562 | 30358504 | ||
U04439.1 | - | 5, 6 | 483 | |||
U16216.1 | - | 6, 7 | 624 | |||
U18867.2 | - | 6, 7 | 628 |
