siRNA Details
Gene Symbol: Utrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011682.4 | NP_035812.3 | 8 | 842 | ||
GenBank mRNA | AK049602.1 | - | 9 | 1199 | ||
AK134010.1 | - | 8 | 836 | |||
AK134424.1 | - | 9 | 1454 | |||
AK163051.1 | - | 8 | 845 | |||
BC011062.1 | - | 9 | 891 | |||
BC050183.1 | - | 9 | 951 | |||
BC062163.1 | - | 9 | 898 | |||
Y12229.1 | - | 8 | 753 |

More Information
Other Species Targeted

Gene Ontology Categories:
