siRNA Details
Gene Symbol: Vcl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009502.4 | NP_033528.3 | 19 | 2918 | ||
GenBank mRNA | AK035184.1 | - | 6 | 949 | ||
AK077850.1 | - | 19 | 2954 | |||
BC008520.1 | - | 19 | 2918 | 3495710 | ||
BC008554.1 | - | 5 | 799 | |||
L18880.1 | - | 19 | 2847 |
