siRNA Details
Gene Symbol: Trio | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081302.1 | NP_001074771.1 | 29 | 4736 | ||
GenBank mRNA | AB106872.1 | - | 28 | 4207 | ||
BC060724.1 | - | 9 | 952 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Trio | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081302.1 | NP_001074771.1 | 29 | 4736 | ||
GenBank mRNA | AB106872.1 | - | 28 | 4207 | ||
BC060724.1 | - | 9 | 952 |