siRNA Details
Gene Symbol: Trio | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081302.1 | NP_001074771.1 | 3 | 671 | ||
GenBank mRNA | AB106872.1 | - | 2 | 142 | ||
BC067059.1 | - | 4 | 480 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Trio | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081302.1 | NP_001074771.1 | 3 | 671 | ||
GenBank mRNA | AB106872.1 | - | 2 | 142 | ||
BC067059.1 | - | 4 | 480 |