siRNA Details
Gene Symbol: Trio | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081302.1 | NP_001074771.1 | 44, 45 | 6747 | ||
GenBank mRNA | BC051169.1 | - | 3, 4 | 214 | ||
BC060724.1 | - | 24, 25 | 2963 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Trio | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081302.1 | NP_001074771.1 | 44, 45 | 6747 | ||
GenBank mRNA | BC051169.1 | - | 3, 4 | 214 | ||
BC060724.1 | - | 24, 25 | 2963 |