siRNA Details
Gene Symbol: 1700021K19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200038.1 | NP_001186967.1 | 7 | 1259 | ||
NM_172615.4 | NP_766203.1 | 7 | 1259 | |||
GenBank mRNA | AK030368.1 | - | 7 | 1233 | ||
AK042428.1 | - | 7 | 1197 | |||
AK122221.1 | - | 7 | 1223 | |||
AK171583.1 | - | 7 | 1243 | |||
BC057307.1 | - | 7 | 1163 | |||
BC060601.1 | - | 7 | 1232 | 5706123 | ||
BC067390.1 | - | 7 | 1244 | 30432960 |
