siRNA Details
Gene Symbol: 1700021K19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200038.1 | NP_001186967.1 | 12 | 1969 | ||
NM_172615.4 | NP_766203.1 | 11 | 1924 | |||
GenBank mRNA | AK030368.1 | - | 11 | 1898 | ||
AK042428.1 | - | 11 | 1862 | |||
AK122221.1 | - | 12 | 1933 | |||
AK136223.1 | - | 4 | 603 | |||
AK170002.1 | - | 12 | 1862 | |||
AK170223.1 | - | 12 | 1820 | |||
AK170312.1 | - | 12 | 2113 | |||
AK171583.1 | - | 11 | 1908 | |||
BC057307.1 | - | 12 | 1873 | |||
BC060601.1 | - | 12 | 1942 | 5706123 | ||
BC067390.1 | - | 11 | 1909 | 30432960 |
