siRNA Details
Gene Symbol: Wnt7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163633.1 | NP_001157105.1 | 2 | 684 | ||
NM_001163634.1 | NP_001157106.1 | 2 | 768 | |||
NM_009528.3 | NP_033554.3 | 2 | 647 | |||
GenBank mRNA | AK087499.1 | - | 2 | 685 | ||
BC052018.1 | - | 2 | 408 | 6401357 | ||
BC058398.1 | - | 2 | 647 | |||
BC066003.1 | - | 2 | 768 | 5701652 | ||
M89802.1 | - | 2 | 276 |
