siRNA Details
Gene Symbol: Ttc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028639.3 | NP_082915.1 | 7 | 1267 | ||
GenBank mRNA | AK040578.1 | - | 7 | 1270 | ||
AK040610.1 | - | 7 | 1238 | |||
AK050263.1 | - | 7 | 1254 | |||
AK080284.1 | - | 7 | 1246 | |||
AK153704.1 | - | 7 | 1249 | |||
BC023773.1 | - | 3 | 225 | |||
BC042512.1 | - | 7 | 944 | 4223801 |
