siRNA Details
Gene Symbol: Rnf165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164504.1 | NP_001157976.1 | 7, 8 | 970 | ||
GenBank mRNA | AK133955.1 | - | 4, 5 | 357 | ||
BB619563.1 | - | 4, 5 | 357 | |||
CF745657.1 | - | 2, 3 | 144 |
