siRNA Details
Gene Symbol: Xpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011728.2 | NP_035858.2 | 4 | 526 | ||
GenBank mRNA | AF354052.1 | - | 2 | 141 | ||
AK008962.1 | - | 4 | 520 | |||
BC029241.1 | - | 503 | 4489063 | |||
BP771655.1 | - | 3 | 473 | |||
CB058423.1 | - | 4 | 528 | |||
X74351.1 | - | 4 | 526 |
