siRNA Details
Gene Symbol: Xpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011728.2 | NP_035858.2 | 6 | 777 | ||
GenBank mRNA | AF354052.1 | - | 4 | 399 | ||
AI838722.1 | - | 1 | 192 | |||
AK133577.1 | - | 1 | 2674 | |||
BP771655.1 | - | 1 | 222 | |||
X74351.1 | - | 6 | 777 |
