siRNA Details
Gene Symbol: Tkfc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145496.1 | NP_663471.1 | 6 | 713 | ||
GenBank mRNA | AK138956.1 | - | 753 | |||
AK171418.1 | - | 7 | 904 | |||
BC021917.1 | - | 6 | 713 | 4481611 |
