siRNA Details
Gene Symbol: BC026585 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033284.1 | NP_001028456.1 | 9 | 944 | ||
GenBank mRNA | AK134211.1 | - | 95 | |||
AK143913.1 | - | 9 | 944 | |||
BC026585.1 | - | 9 | 946 | |||
BC045198.1 | - | 9 | 941 | 5065108 | ||
BC057972.1 | - | 3 | 446 |
