siRNA Details
Gene Symbol: Rcsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038846.1 | NP_001033935.1 | 4 | 526 | ||
NM_178593.3 | NP_848708.2 | 5 | 616 | |||
GenBank mRNA | AK133961.1 | - | 5 | 571 | ||
AK158429.1 | - | 5 | 557 | |||
AK168816.1 | - | 4 | 497 | |||
AK171323.1 | - | 506 | ||||
AK171914.1 | - | 5 | 539 | |||
BC025872.1 | - | 5 | 536 | 5037953 |
