siRNA Details
Gene Symbol: AI607873 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204910.1 | NP_001191839.1 | 5 | 1955 | ||
GenBank mRNA | BC150711.1 | - | 1844 | 9087622 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: AI607873 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204910.1 | NP_001191839.1 | 5 | 1955 | ||
GenBank mRNA | BC150711.1 | - | 1844 | 9087622 |