siRNA Details
Gene Symbol: Cdc42bpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033285.1 | NP_001028457.1 | 11 | 1476 | ||
GenBank mRNA | AK138649.1 | - | 12 | 1965 | ||
BC158017.1 | - | 11 | 1476 | 9007323 | ||
BC158095.1 | - | 11 | 1476 | 9007382 |
