siRNA Details
Gene Symbol: Slc30a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033286.2 | NP_001028458.1 | 1 | 332 | ||
GenBank mRNA | AK053537.1 | - | 1 | 333 | ||
AK136932.1 | - | 1 | 140 | |||
BB665136.1 | - | 1 | 333 | |||
BC054548.1 | - | 1 | 290 |
Gene Symbol: Gm2061 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK138296.1 | - | 1 | 387 |
