siRNA Details
Gene Symbol: Kcnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK033953.1 | - | 18 | 1813 | ||
AK039085.1 | - | 2 | 214 | |||
AK129355.1 | - | 12 | 1283 | |||
AK147625.1 | - | 16 | 3762 | |||
BC141190.1 | - | 17 | 1858 | 9055802 | ||
BC171963.1 | - | 17 | 1858 | 9054249 | ||
FJ628361.1 | - | 18 | 1856 |
