siRNA Details
Gene Symbol: Kcnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK033953.1 | - | 24 | 2772 | ||
AK039085.1 | - | 8 | 1174 | |||
AK046816.1 | - | 13 | 1664 | |||
AK048406.1 | - | 6 | 650 | |||
AK129355.1 | - | 18 | 2242 | |||
AK147625.1 | - | 22 | 4721 | |||
BC057073.1 | - | 1 | 71 | |||
BC141190.1 | - | 23 | 2817 | 9055802 | ||
BC171963.1 | - | 23 | 2817 | 9054249 | ||
FJ628361.1 | - | 24 | 2815 |
