siRNA Details
Gene Symbol: Kcnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK033953.1 | - | 32 | 3684 | ||
AK039085.1 | - | 15 | 3048 | |||
AK046816.1 | - | 20 | 2780 | |||
AK048406.1 | - | 14 | 1562 | |||
AK081253.1 | - | 4 | 309 | |||
AK129355.1 | - | 26 | 3154 | |||
AK147625.1 | - | 30 | 5633 | |||
BC057073.1 | - | 8 | 1270 | |||
BC141190.1 | - | 31 | 3729 | 9055802 | ||
BC171963.1 | - | 31 | 3729 | 9054249 | ||
FJ628361.1 | - | 32 | 3727 |
