siRNA Details
Gene Symbol: BC005624 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144885.2 | NP_659134.1 | 8 | 829 | ||
GenBank mRNA | AK158706.1 | - | 794 | |||
AK163554.1 | - | 802 | ||||
AK168109.1 | - | 8 | 749 | |||
BC005624.1 | - | 8 | 774 | 3962182 |
