siRNA Details
Gene Symbol: Rabepk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145522.4 | NP_663497.2 | 5 | 612 | ||
GenBank mRNA | AK018405.1 | - | 1 | 906 | ||
AK035343.1 | - | 5 | 492 | |||
AK153022.1 | - | 5 | 516 | |||
BC019800.1 | - | 5 | 488 | 5150243 |
