siRNA Details
Gene Symbol: Tnks1bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081260.2 | NP_001074729.1 | 6 | 2890 | ||
GenBank mRNA | AK129436.1 | - | 2 | 1583 | ||
BC025943.1 | - | 1 | 339 | |||
BC058639.1 | - | 2 | 1942 | |||
BC150947.1 | - | 6 | 2810 | 9087858 |
