siRNA Details
Gene Symbol: Bpifb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199303.2 | NP_955007.2 | 14, 15 | 1333 | ||
GenBank mRNA | AK087711.1 | - | 3, 4 | 187 | ||
BC048083.1 | - | 14, 15 | 1333 | 6312228 |
