siRNA Details
Gene Symbol: Otud7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025613.1 | NP_001020784.1 | 13 | 2363 | ||
NM_001025614.1 | NP_001020785.1 | 12 | 2305 | |||
GenBank mRNA | BC141397.1 | - | 12 | 2195 | 9056010 | |
BC141398.1 | - | 12 | 2195 | 9056011 |
