siRNA Details
Gene Symbol: Eif4g3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256195.1 | NP_001243124.1 | 31 | 4695 | ||
NM_001256198.1 | NP_001243127.1 | 31 | 4752 | |||
NM_172703.3 | NP_766291.2 | 31 | 4599 | |||
GenBank mRNA | AK029440.1 | - | 14 | 1679 | ||
AK133389.1 | - | 11 | 1163 | |||
BC022766.1 | - | 7 | 802 | |||
BC023898.1 | - | 18 | 2441 | |||
BC047531.1 | - | 20 | 2823 | |||
BC048848.1 | - | 10 | 1105 | |||
BC057913.1 | - | 11 | 1221 | |||
BC072600.1 | - | 31 | 4585 | 6410320 | ||
BC075705.1 | - | 16 | 2165 | |||
BC079885.1 | - | 31 | 4649 | |||
BC082549.1 | - | 31 | 4716 |

More Information
Other Species Targeted

Gene Ontology Categories:
