siRNA Details
Gene Symbol: Sh2d5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099631.1 | NP_001093101.1 | 5 | 438 | ||
GenBank mRNA | AK032598.1 | - | 5 | 372 | ||
BC036961.1 | - | 6 | 472 | |||
DQ832275.1 | - | 5 | 438 |
