siRNA Details
Gene Symbol: Ssh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198109.4 | NP_932777.2 | 10 | 876 | ||
GenBank mRNA | AB099287.1 | - | 10 | 911 | ||
AK089308.1 | - | 10 | 880 | |||
AK155557.1 | - | 10 | 901 | |||
AK165380.1 | - | 9 | 841 | |||
AK170787.1 | - | 9 | 801 | |||
AK171210.1 | - | 10 | 925 | |||
AK171556.1 | - | 10 | 891 | |||
AK173144.1 | - | 9 | 832 | |||
BC046529.1 | - | 4 | 358 |
