siRNA Details
Gene Symbol: Myo1h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164573.1 | NP_001158045.1 | 29, 30 | 2948 | ||
GenBank mRNA | AK014505.1 | - | 7, 8 | 991 | ||
BC144867.1 | - | 8, 9 | 853 | 9053415 | ||
BC144868.1 | - | 8, 9 | 881 | |||
BC144869.1 | - | 8, 9 | 781 | |||
BC144870.1 | - | 8, 9 | 829 | 9053418 | ||
BC144872.1 | - | 6, 7 | 910 |
