siRNA Details
Gene Symbol: Tnrc18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122730.2 | NP_001116202.2 | 11 | 4496 | ||
NM_178242.2 | NP_839973.1 | 11 | 4496 | |||
GenBank mRNA | AK141849.1 | - | 8 | 3594 | ||
AK173280.1 | - | 8 | 2254 | |||
BC025631.1 | - | 6 | 2029 | |||
BC037158.1 | - | 1 | 92 | |||
BC049818.1 | - | 11 | 4496 | 6511384 |
