siRNA Details
Gene Symbol: Fkbp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153573.1 | NP_705801.1 | 1 | 192 | ||
GenBank mRNA | AK030620.1 | - | 1 | 242 | ||
AK044876.1 | - | 1 | 228 | |||
AK081692.1 | - | 1 | 219 | |||
BC024846.1 | - | 529 | ||||
BC029109.1 | - | 1 | 192 | 3985764 | ||
BC034313.1 | - | 1 | 217 | 5365046 |
