siRNA Details
Gene Symbol: Iqsec1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134383.1 | NP_001127855.1 | 11 | 2759 | ||
NM_001134384.1 | NP_001127856.1 | 11 | 2793 | |||
GenBank mRNA | AK220355.1 | - | 9 | 1210 | ||
BC026481.1 | - | 9 | 1263 |
