siRNA Details
Gene Symbol: Mgam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171003.1 | NP_001164474.1 | 16 | 1904 | ||
GenBank mRNA | EU937529.1 | - | 16 | 1904 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mgam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171003.1 | NP_001164474.1 | 16 | 1904 | ||
GenBank mRNA | EU937529.1 | - | 16 | 1904 |