siRNA Details
Gene Symbol: Mgam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171003.1 | NP_001164474.1 | 5 | 581 | ||
GenBank mRNA | AY229885.1 | - | 581 | |||
AY229886.1 | - | 457 | ||||
EU937529.1 | - | 5 | 581 |
