siRNA Details
Gene Symbol: Shisa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290291.1 | NP_001277220.1 | 5 | 1333 | ||
NM_172737.4 | NP_766325.3 | 6 | 1384 | |||
GenBank mRNA | AK085127.1 | - | 5 | 606 | ||
AK145016.1 | - | 4 | 618 | |||
BY734785.1 | - | 4 | 618 |
