siRNA Details
Gene Symbol: Zbtb45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024699.1 | NP_001019870.1 | 2 | 370 | ||
GenBank mRNA | AK154877.1 | - | 2 | 446 | ||
AK155880.1 | - | 2 | 427 | |||
AK166382.1 | - | 439 | ||||
BC094359.1 | - | 2 | 370 | 6828341 |
