siRNA Details
Gene Symbol: Atp1a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290469.1 | NP_001277398.1 | 15 | 2181 | ||
GenBank mRNA | BC020177.1 | - | 15 | 2139 | 4506376 | |
BC027000.1 | - | 8 | 1201 | |||
BC027114.1 | - | 3 | 310 | |||
BC034645.1 | - | 15 | 2137 | 5357731 | ||
BC037206.1 | - | 15 | 2136 | 5362423 | ||
BC042894.1 | - | 15 | 2181 | 5364812 |
